siRNA Details
| Gene Symbol: Evi5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007964.2 | NP_031990.2 | 18 | 2475 | ||
| GenBank mRNA | AK136326.1 | - | 8 | 1256 | ||
| AK164208.1 | - | 4 | 583 | |||
| CN456828.1 | - | 5 | 656 | |||
| U53586.1 | - | 18 | 2475 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Evi5| Gene Symbol: Evi5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007964.2 | NP_031990.2 | 18 | 2475 | ||
| GenBank mRNA | AK136326.1 | - | 8 | 1256 | ||
| AK164208.1 | - | 4 | 583 | |||
| CN456828.1 | - | 5 | 656 | |||
| U53586.1 | - | 18 | 2475 | |||