siRNA Details
| Gene Symbol: Ext1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010162.2 | NP_034292.2 | 1 | 938 | ||
| GenBank mRNA | AK132327.1 | - | 1 | 915 | ||
| BC004741.1 | - | 1 | 302 | 3499743 | ||
| U78539.1 | - | 1 | 442 | |||
| X96639.1 | - | 1 | 712 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ext1| Gene Symbol: Ext1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010162.2 | NP_034292.2 | 1 | 938 | ||
| GenBank mRNA | AK132327.1 | - | 1 | 915 | ||
| BC004741.1 | - | 1 | 302 | 3499743 | ||
| U78539.1 | - | 1 | 442 | |||
| X96639.1 | - | 1 | 712 | |||