siRNA Details
| Gene Symbol: Man2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172903.4 | NP_766491.2 | 5 | 985 | ||
| GenBank mRNA | AK029913.1 | - | 5 | 796 | ||
| AK044028.1 | - | 4 | 1723 | |||
| AK161799.1 | - | 4 | 644 | |||
| DQ631806.1 | - | 3 | 500 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Man2a2| Gene Symbol: Man2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172903.4 | NP_766491.2 | 5 | 985 | ||
| GenBank mRNA | AK029913.1 | - | 5 | 796 | ||
| AK044028.1 | - | 4 | 1723 | |||
| AK161799.1 | - | 4 | 644 | |||
| DQ631806.1 | - | 3 | 500 | |||