siRNA Details
| Gene Symbol: Man2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172903.4 | NP_766491.2 | 11 | 1910 | ||
| GenBank mRNA | AK029913.1 | - | 11 | 1721 | ||
| AK044028.1 | - | 10 | 2648 | |||
| AK044561.1 | - | 1 | 48 | |||
| AK161799.1 | - | 10 | 1569 | |||
| DQ631806.1 | - | 9 | 1425 | |||

Man2a2
Genomic Map