siRNA Details
| Gene Symbol: Eya1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252192.1 | NP_001239121.1 | 14 | 1895 | ||
| NM_010164.2 | NP_034294.2 | 15 | 2011 | |||
| GenBank mRNA | AJ007995.1 | - | 2 | 125 | ||
| AK042412.1 | - | 14 | 1818 | |||
| AK153788.1 | - | 15 | 2017 | |||
| AK162111.1 | - | 7 | 752 | |||
| BC060260.1 | - | 15 | 2011 | 6848408 | ||
| BC066860.1 | - | 15 | 1665 | 30089692 | ||
| U61110.1 | - | 14 | 1568 | |||

Eya1
Genomic Map