siRNA Details
| Gene Symbol: Atp6v0a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080467.3 | NP_536715.3 | 20 | 2585 | ||
| GenBank mRNA | AB050903.1 | - | 19 | 2311 | ||
| AF326316.1 | - | 20 | 2379 | |||
| AF435090.1 | - | 20 | 2573 | |||
| BC046979.1 | - | 21 | 2607 | 6315485 | ||

Atp6v0a4
Genomic Map