siRNA Details
| Gene Symbol: Atp6v0a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080467.3 | NP_536715.3 | 16 | 1992 | ||
| GenBank mRNA | AB050903.1 | - | 15 | 1718 | ||
| AF326316.1 | - | 16 | 1786 | |||
| AF435090.1 | - | 16 | 1980 | |||
| BC046979.1 | - | 17 | 2014 | 6315485 | ||

Atp6v0a4
Genomic Map