siRNA Details
| Gene Symbol: Eya2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271962.1 | NP_001258891.1 | 2, 3 | 160 | ||
| NM_001271963.1 | NP_001258892.1 | 2, 3 | 336 | |||
| NM_010165.3 | NP_034295.1 | 2, 3 | 337 | |||
| GenBank mRNA | AK137718.1 | - | 2, 3 | 332 | ||
| AK141765.1 | - | 2, 3 | 336 | |||
| AY212511.1 | - | 1, 2 | 97 | |||
| AY212512.1 | - | 1, 2 | 97 | |||
| BC003755.1 | - | 2, 3 | 337 | 3497245 | ||
| CF724561.1 | - | 1, 2 | 36 | |||
| U61111.1 | - | 1, 2 | 78 | |||
| U71208.1 | - | 2, 3 | 162 | |||
| U81603.1 | - | 1, 2 | 77 | |||

Eya2
Genomic Map