siRNA Details
| Gene Symbol: Eya3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010166.3 | NP_034296.2 | 16 | 1563 | ||
| NM_210071.2 | NP_997592.1 | 15 | 1524 | |||
| NM_211357.2 | NP_997596.1 | 13 | 1343 | |||
| GenBank mRNA | AJ007996.1 | - | 3 | 256 | ||
| AK144653.1 | - | 15 | 1515 | |||
| BC052860.1 | - | 14 | 1327 | |||
| BC063259.1 | - | 15 | 1452 | 6406090 | ||
| U61112.1 | - | 12 | 1228 | |||
| U81604.1 | - | 16 | 1531 | |||

Eya3
Genomic Map