siRNA Details
| Gene Symbol: Eya3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010166.3 | NP_034296.2 | 14 | 1319 | ||
| NM_210071.2 | NP_997592.1 | 13 | 1280 | |||
| NM_211357.2 | NP_997596.1 | 11 | 1099 | |||
| GenBank mRNA | AJ007996.1 | - | 1 | 12 | ||
| AK144653.1 | - | 13 | 1271 | |||
| BC052860.1 | - | 12 | 1083 | |||
| BC063259.1 | - | 13 | 1208 | 6406090 | ||
| U61112.1 | - | 10 | 984 | |||
| U81604.1 | - | 14 | 1287 | |||

Eya3
Genomic Map