siRNA Details
| Gene Symbol: Ezh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146689.1 | NP_001140161.1 | 19 | 2255 | ||
| NM_007971.2 | NP_031997.2 | 19 | 2267 | |||
| GenBank mRNA | AK157853.1 | - | 19 | 2268 | ||
| AK220174.1 | - | 19 | 2222 | |||
| BC003772.1 | - | 19 | 2176 | 3492110 | ||
| BC016391.1 | - | 19 | 2189 | 3586689 | ||
| BC079538.1 | - | 19 | 2206 | 6817366 | ||
| U52951.1 | - | 19 | 2257 | |||

Ezh2
Genomic Map