siRNA Details
| Gene Symbol: Elmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080288.2 | NP_525027.1 | 19, 20 | 2166 | ||
| NM_198093.3 | NP_932761.1 | 4, 5 | 689 | |||
| GenBank mRNA | AF398883.1 | - | 18, 19 | 1830 | ||
| AK032085.1 | - | 4, 5 | 599 | |||
| AK053306.1 | - | 4, 5 | 600 | |||
| AK220253.1 | - | 19, 20 | 2054 | |||
| BC024727.1 | - | 4, 5 | 586 | |||
| BC031782.1 | - | 8, 9 | 904 | 3990906 | ||

Elmo1
Genomic Map