siRNA Details
Gene Symbol: Elmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080288.2 | NP_525027.1 | 22 | 2370 | ||
NM_198093.3 | NP_932761.1 | 7 | 893 | |||
GenBank mRNA | AF398883.1 | - | 21 | 2034 | ||
AK220253.1 | - | 22 | 2258 | |||
BC024727.1 | - | 7 | 790 | |||
BC031782.1 | - | 11 | 1108 | 3990906 |
