siRNA Details
Gene Symbol: Ube4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145400.3 | NP_663375.3 | 9, 10 | 1376 | ||
GenBank mRNA | AK028850.1 | - | 9 | 2900 | ||
AK172893.1 | - | 9, 10 | 1336 | |||
BC006649.1 | - | 10, 11 | 1515 | 3498834 |
