siRNA Details
| Gene Symbol: F5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137521.1 | - | 3 | 421 | ||
| AK137537.1 | - | 3 | 398 | |||
| AK139240.1 | - | 3 | 415 | |||
| BY722729.1 | - | 3 | 395 | |||
| U52925.1 | - | 3 | 296 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F5| Gene Symbol: F5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137521.1 | - | 3 | 421 | ||
| AK137537.1 | - | 3 | 398 | |||
| AK139240.1 | - | 3 | 415 | |||
| BY722729.1 | - | 3 | 395 | |||
| U52925.1 | - | 3 | 296 | |||