siRNA Details
| Gene Symbol: F7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089402.1 | - | 8 | 1206 | ||
| AK154742.1 | - | 8 | 1206 | |||
| AK155323.1 | - | 6 | 554 | |||
| AK171362.1 | - | 8 | 1203 | |||
| BC061149.1 | - | 8 | 1189 | 30305571 | ||
| U44795.1 | - | 8 | 1195 | |||

F7
Genomic Map