siRNA Details
| Gene Symbol: F7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089402.1 | - | 8 | 833 | ||
| AK154742.1 | - | 8 | 833 | |||
| AK171362.1 | - | 8 | 830 | |||
| BC061149.1 | - | 8 | 816 | 30305571 | ||
| U44795.1 | - | 8 | 822 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F7| Gene Symbol: F7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089402.1 | - | 8 | 833 | ||
| AK154742.1 | - | 8 | 833 | |||
| AK171362.1 | - | 8 | 830 | |||
| BC061149.1 | - | 8 | 816 | 30305571 | ||
| U44795.1 | - | 8 | 822 | |||