siRNA Details
Gene Symbol: Bmp2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080708.1 | NP_542439.1 | 15 | 2175 | ||
GenBank mRNA | AY050249.1 | - | 15 | 2175 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bmp2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080708.1 | NP_542439.1 | 15 | 2175 | ||
GenBank mRNA | AY050249.1 | - | 15 | 2175 |