siRNA Details
| Gene Symbol: Acsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004897.1 | - | 3 | 315 | ||
| AK145900.1 | - | 3 | 442 | |||
| AK149406.1 | - | 3 | 320 | |||
| AK152772.1 | - | 3 | 320 | |||
| AK153050.1 | - | 3 | 448 | |||
| AK158388.1 | - | 3 | 399 | |||
| AK161189.1 | - | 3 | 294 | |||
| AK168078.1 | - | 3 | 320 | |||
| BC056644.1 | - | 3 | 304 | 5098645 | ||
| BY094311.1 | - | 3 | 322 | |||
| U15977.1 | - | 2 | 233 | |||

Acsl1
Genomic Map