siRNA Details
| Gene Symbol: Faf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007983.2 | NP_032009.2 | 4 | 588 | ||
| GenBank mRNA | AK053752.1 | - | 4 | 523 | ||
| AK183710.1 | - | 3 | 153 | |||
| AK186332.1 | - | 3 | 153 | |||
| BC065098.1 | - | 4 | 588 | 30362116 | ||
| U39643.1 | - | 4 | 598 | |||

Faf1
Genomic Map