siRNA Details
| Gene Symbol: Faf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007983.2 | NP_032009.2 | 13, 14 | 1664 | ||
| GenBank mRNA | AK053752.1 | - | 13, 14 | 1599 | ||
| BC065098.1 | - | 13, 14 | 1664 | 30362116 | ||
| U39643.1 | - | 13, 14 | 1674 | |||

Faf1
Genomic Map