siRNA Details
| Gene Symbol: Nek8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080849.3 | NP_543125.1 | 10 | 1457 | ||
| GenBank mRNA | AF407579.1 | - | 10 | 1407 | ||
| AK014546.1 | - | 8 | 1633 | |||
| AK154358.1 | - | 10 | 1420 | |||
| BC057995.1 | - | 9 | 1539 | |||
| BC070457.1 | - | 10 | 1406 | 30603080 | ||

Nek8
Genomic Map