siRNA Details
| Gene Symbol: Nek8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080849.3 | NP_543125.1 | 11 | 1577 | ||
| GenBank mRNA | AF407579.1 | - | 11 | 1527 | ||
| AK014546.1 | - | 9 | 1753 | |||
| AK154358.1 | - | 11 | 1540 | |||
| BC057995.1 | - | 10 | 1659 | |||
| BC070457.1 | - | 11 | 1526 | 30603080 | ||

Nek8
Genomic Map