siRNA Details
| Gene Symbol: Fah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010176.4 | NP_034306.2 | 11 | 1163 | ||
| GenBank mRNA | AI848212.1 | - | 4 | 431 | ||
| AK143759.1 | - | 11 | 992 | |||
| AK158808.1 | - | 11 | 1049 | |||
| BC010767.1 | - | 11 | 948 | 4242421 | ||
| M84145.1 | - | 11 | 989 | |||
| Z11774.1 | - | 11 | 975 | |||

Fah
Genomic Map