siRNA Details
| Gene Symbol: Fah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010176.4 | NP_034306.2 | 2 | 390 | ||
| GenBank mRNA | AK143759.1 | - | 2 | 219 | ||
| AK158808.1 | - | 2 | 276 | |||
| BC010767.1 | - | 2 | 175 | 4242421 | ||
| M84145.1 | - | 2 | 216 | |||
| Z11774.1 | - | 2 | 202 | |||

Fah
Genomic Map