siRNA Details
| Gene Symbol: Fanca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016925.3 | NP_058621.2 | 32 | 3093 | ||
| GenBank mRNA | AF178934.1 | - | 32 | 3089 | ||
| AF208116.1 | - | 32 | 3095 | |||
| AF208120.1 | - | 11 | 1176 | |||
| BC150705.1 | - | 32 | 3091 | 9087616 | ||

Fanca
Genomic Map