siRNA Details
| Gene Symbol: Fancc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042673.2 | NP_001036138.1 | 4 | 613 | ||
| NM_001282942.1 | NP_001269871.1 | 3 | 416 | |||
| NM_007985.3 | NP_032011.2 | 4 | 613 | |||
| GenBank mRNA | AK035488.1 | - | 4 | 367 | ||
| AK042226.1 | - | 4 | 583 | |||
| AK080697.1 | - | 594 | ||||
| AK082717.1 | - | 3 | 394 | |||
| AK147025.1 | - | 4 | 593 | |||
| AK163161.1 | - | 4 | 614 | |||
| L08266.1 | - | 4 | 594 | |||

Fancc
Genomic Map