siRNA Details
| Gene Symbol: Fap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007986.3 | NP_032012.1 | 19 | 1753 | ||
| GenBank mRNA | BC019190.1 | - | 19 | 1657 | 5028579 | |
| Y10007.1 | - | 19 | 1753 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fap| Gene Symbol: Fap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007986.3 | NP_032012.1 | 19 | 1753 | ||
| GenBank mRNA | BC019190.1 | - | 19 | 1657 | 5028579 | |
| Y10007.1 | - | 19 | 1753 | |||