siRNA Details
| Gene Symbol: Slc7a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080852.2 | NP_543128.1 | 1 | 324 | ||
| GenBank mRNA | AB055005.1 | - | 1 | 329 | ||
| AJ417663.1 | - | 1 | 286 | |||
| AK085644.1 | - | 1 | 313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc7a12| Gene Symbol: Slc7a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080852.2 | NP_543128.1 | 1 | 324 | ||
| GenBank mRNA | AB055005.1 | - | 1 | 329 | ||
| AJ417663.1 | - | 1 | 286 | |||
| AK085644.1 | - | 1 | 313 | |||