siRNA Details
| Gene Symbol: Fem1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010193.4 | NP_034323.1 | 2 | 1245 | ||
| GenBank mRNA | AB022863.1 | - | 2 | 1155 | ||
| AF064448.1 | - | 2 | 976 | |||
| AF178633.1 | - | 2 | 902 | |||
| AK032338.1 | - | 2 | 1222 | |||
| AK122272.1 | - | 1 | 443 | |||
| AK132692.1 | - | 2 | 1238 | |||
| AK145371.1 | - | 1 | 608 | |||
| AK149329.1 | - | 2 | 1208 | |||
| AK154060.1 | - | 2 | 1533 | |||
| BC068236.1 | - | 2 | 1199 | 30544277 | ||

Fem1b
Genomic Map