siRNA Details
| Gene Symbol: Fer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037997.3 | NP_001033086.2 | 15 | 2000 | ||
| NM_001286415.1 | NP_001273344.1 | 14 | 1828 | |||
| NM_008000.2 | NP_032026.2 | 10 | 1282 | |||
| GenBank mRNA | AK082799.1 | - | 12 | 1493 | ||
| AK157828.1 | - | 15 | 1964 | |||
| BC051249.1 | - | 7 | 744 | |||
| BC058100.1 | - | 14 | 1828 | 6813507 | ||
| M32054.1 | - | 10 | 1282 | |||
| U76762.1 | - | 14 | 1933 | |||

Fer
Genomic Map