siRNA Details
| Gene Symbol: Fgd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008001.4 | NP_032027.2 | 4 | 1462 | ||
| GenBank mRNA | AK148151.1 | - | 4 | 1463 | ||
| AK162865.1 | - | 2 | 452 | |||
| BC011462.1 | - | 4 | 1399 | 3674293 | ||
| CN531569.1 | - | 3 | 575 | |||
| U22325.1 | - | 4 | 1061 | |||

Fgd1
Genomic Map