siRNA Details
| Gene Symbol: Fgf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290414.1 | NP_001277343.1 | 7 | 1065 | ||
| NM_001290415.1 | NP_001277344.1 | 5 | 576 | |||
| NM_010200.3 | NP_034330.2 | 5 | 1060 | |||
| GenBank mRNA | AF020737.1 | - | 5 | 693 | ||
| AK141848.1 | - | 5 | 576 | |||
| BC018238.1 | - | 5 | 1060 | 4481123 | ||
| U66202.1 | - | 5 | 687 | |||

Fgf13
Genomic Map