siRNA Details
| Gene Symbol: Fgfr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163215.2 | NP_001156687.1 | 5 | 788 | ||
| NM_001163216.2 | NP_001156688.1 | 3 | 503 | |||
| NM_001163217.2 | NP_001156689.1 | 4 | 557 | |||
| NM_001205270.1 | NP_001192199.1 | 5 | 762 | |||
| NM_008010.5 | NP_032036.2 | 5 | 762 | |||
| GenBank mRNA | AF024638.1 | - | 3 | 411 | ||
| AK140217.1 | - | 4 | 679 | |||
| AK143592.1 | - | 5 | 695 | |||
| AK163495.1 | - | 3 | 341 | |||
| AK163547.1 | - | 4 | 679 | |||
| BC053056.1 | - | 5 | 672 | 5708838 | ||
| CB247234.1 | - | 5 | 633 | |||
| M81342.1 | - | 5 | 680 | |||
| S56291.1 | - | 4 | 507 | |||
| X58255.1 | - | 5 | 666 | |||

Fgfr3
Genomic Map