siRNA Details
| Gene Symbol: Fhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010213.3 | NP_034343.2 | 2 | 216 | ||
| GenBank mRNA | AF114382.1 | - | 2 | 187 | ||
| AF134772.1 | - | 2 | 213 | |||
| AF149826.1 | - | 1 | 14 | |||
| AI426098.1 | - | 2 | 213 | |||
| BC144903.1 | - | 2 | 180 | 9053451 | ||
| BC145939.1 | - | 2 | 180 | 40131242 | ||

Fhl3
Genomic Map