siRNA Details
| Gene Symbol: Smc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301412.1 | NP_001288341.1 | 22, 23 | 3363 | ||
| NM_008017.4 | NP_032043.3 | 22, 23 | 3287 | |||
| GenBank mRNA | AJ534939.1 | - | 21, 22 | 3118 | ||
| AK145341.1 | - | 22, 23 | 3317 | |||
| BC071232.1 | - | 7, 8 | 1022 | |||
| BC094380.1 | - | 22, 23 | 3259 | 30543190 | ||
| U42385.1 | - | 5, 6 | 730 | |||

Smc2
Genomic Map