siRNA Details
| Gene Symbol: Sh3pxd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164717.1 | NP_001158189.1 | 14 | 2002 | ||
| NM_008018.4 | NP_032044.2 | 15 | 2086 | |||
| GenBank mRNA | AJ007012.1 | - | 15 | 2086 | ||
| AK085227.1 | - | 8 | 1925 | |||
| BC118022.1 | - | 14 | 1838 | 40097448 | ||

Sh3pxd2a
Genomic Map