siRNA Details
| Gene Symbol: Fkbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010219.3 | NP_034349.1 | 6 | 856 | ||
| GenBank mRNA | AK051433.1 | - | 882 | |||
| AK083912.1 | - | 6 | 856 | |||
| AK089972.1 | - | 6 | 865 | |||
| AK159211.1 | - | 6 | 883 | |||
| AK160202.1 | - | 6 | 805 | |||
| BC003447.1 | - | 6 | 873 | 2651490 | ||
| X17069.2 | - | 6 | 722 | |||
| X70887.1 | - | 6 | 753 | |||

Fkbp4
Genomic Map