siRNA Details
| Gene Symbol: Fkbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010219.3 | NP_034349.1 | 10 | 1429 | ||
| GenBank mRNA | AK035395.1 | - | 2 | 119 | ||
| AK051433.1 | - | 1548 | ||||
| AK083912.1 | - | 10 | 1429 | |||
| AK089972.1 | - | 10 | 1438 | |||
| AK159211.1 | - | 10 | 1371 | |||
| AK160202.1 | - | 10 | 1378 | |||
| BC003447.1 | - | 10 | 1446 | 2651490 | ||
| X17069.2 | - | 10 | 1295 | |||
| X70887.1 | - | 10 | 1326 | |||

Fkbp4
Genomic Map