siRNA Details
| Gene Symbol: Fkbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010220.4 | NP_034350.1 | 5 | 649 | ||
| GenBank mRNA | AK169795.1 | - | 5 | 590 | ||
| BC015260.1 | - | 5 | 588 | 4237766 | ||
| BU613862.1 | - | 3 | 316 | |||
| CA317455.1 | - | 5 | 612 | |||
| CT010372.1 | - | 4 | 430 | |||
| U16959.1 | - | 5 | 649 | |||
| U36220.1 | - | 5 | 577 | |||

Fkbp5
Genomic Map