siRNA Details
| Gene Symbol: Foxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008021.4 | NP_032047.4 | 8 | 1573 | ||
| GenBank mRNA | AK049177.1 | - | 8 | 1514 | ||
| BC006788.1 | - | 1 | 17 | |||
| BC065067.1 | - | 8 | 1573 | 6417437 | ||
| BC075723.1 | - | 8 | 1592 | 6515008 | ||
| Y11245.1 | - | 8 | 1481 | |||

Foxm1
Genomic Map