siRNA Details
| Gene Symbol: Foxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008021.4 | NP_032047.4 | 2 | 417 | ||
| GenBank mRNA | AK041638.1 | - | 2 | 385 | ||
| AK049177.1 | - | 2 | 358 | |||
| BC065067.1 | - | 2 | 417 | 6417437 | ||
| BC075723.1 | - | 2 | 436 | 6515008 | ||
| Y11245.1 | - | 2 | 325 | |||

Foxm1
Genomic Map