siRNA Details
Gene Symbol: Flot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040403.1 | NP_001035493.1 | 5 | 573 | ||
NM_001284227.1 | NP_001271156.1 | 6 | 657 | |||
NM_001284228.1 | NP_001271157.1 | 6 | 520 | |||
NM_008028.2 | NP_032054.1 | 6 | 664 | |||
GenBank mRNA | AK004589.1 | - | 5 | 511 | ||
AK030199.1 | - | 5 | 536 | |||
AK149530.1 | - | 6 | 595 | |||
AK170557.1 | - | 5 | 574 | |||
AK188583.1 | - | 1 | 17 | |||
BC070423.1 | - | 6 | 622 | 6403816 | ||
U07890.1 | - | 6 | 534 |
