siRNA Details
| Gene Symbol: Flt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010228.3 | NP_034358.2 | 3 | 625 | ||
| GenBank mRNA | AK005502.1 | - | 3 | 648 | ||
| AK146906.1 | - | 3 | 650 | |||
| AK167497.1 | - | 3 | 432 | |||
| AK167596.1 | - | 3 | 649 | |||
| BC029674.1 | - | 3 | 449 | 5368921 | ||
| D88689.1 | - | 3 | 622 | |||
| D88690.1 | - | 3 | 622 | |||
| L07297.1 | - | 3 | 397 | |||
| X78568.1 | - | 3 | 619 | |||

Flt1
Genomic Map