siRNA Details
| Gene Symbol: Flt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010228.3 | NP_034358.2 | 5 | 834 | ||
| GenBank mRNA | AK005502.1 | - | 5 | 857 | ||
| AK146906.1 | - | 5 | 859 | |||
| AK167497.1 | - | 5 | 641 | |||
| AK167596.1 | - | 5 | 858 | |||
| BC029674.1 | - | 5 | 658 | 5368921 | ||
| D88689.1 | - | 5 | 831 | |||
| D88690.1 | - | 5 | 831 | |||
| L07297.1 | - | 5 | 606 | |||
| X78568.1 | - | 5 | 825 | |||

Flt1
Genomic Map