siRNA Details
| Gene Symbol: Flt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010229.2 | NP_034359.2 | 12 | 1721 | ||
| GenBank mRNA | AK149292.1 | - | 12 | 1539 | ||
| BC109003.1 | - | 12 | 1514 | |||
| BC109004.1 | - | 12 | 1526 | |||
| M64689.1 | - | 12 | 1527 | |||
| X59398.1 | - | 12 | 1578 | |||

Flt3
Genomic Map