siRNA Details
| Gene Symbol: Fmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008030.1 | NP_032056.1 | 9 | 1320 | ||
| GenBank mRNA | AK149440.1 | - | 9 | 1347 | ||
| U87147.1 | - | 9 | 1320 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fmo3| Gene Symbol: Fmo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008030.1 | NP_032056.1 | 9 | 1320 | ||
| GenBank mRNA | AK149440.1 | - | 9 | 1347 | ||
| U87147.1 | - | 9 | 1320 | |||