siRNA Details
| Gene Symbol: Aff2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008032.3 | NP_032058.2 | 3 | 1447 | ||
| GenBank mRNA | AJ001549.1 | - | 3 | 1451 | ||
| AJ748649.1 | - | 1 | 602 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aff2| Gene Symbol: Aff2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008032.3 | NP_032058.2 | 3 | 1447 | ||
| GenBank mRNA | AJ001549.1 | - | 3 | 1451 | ||
| AJ748649.1 | - | 1 | 602 | |||