siRNA Details
| Gene Symbol: Asb13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178283.4 | NP_840068.1 | 5 | 664 | ||
| GenBank mRNA | AF403041.1 | - | 5 | 664 | ||
| AK078289.1 | - | 5 | 620 | |||
| AK133892.1 | - | 5 | 588 | |||
| BC018240.1 | - | 5 | 618 | 4239996 | ||

Asb13
Genomic Map