siRNA Details
Gene Symbol: Srgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081011.2 | NP_001074480.2 | 8 | 1646 | ||
GenBank mRNA | BC030457.1 | - | 7 | 953 | ||
BC151081.1 | - | 7 | 1264 | 9087994 | ||
BC151082.1 | - | 7 | 1264 | 9087995 | ||
BC158055.1 | - | 7 | 1262 | 9088163 | ||
BC172152.1 | - | 7 | 1264 | 9054822 | ||
BE653295.1 | - | 1 | 69 |
